siRNA Details
Gene Symbol: Epn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252188.1 | NP_001239117.1 | 9 | 1719 | ||
NM_001252189.1 | NP_001239118.1 | 7 | 1475 | |||
NM_010148.3 | NP_034278.1 | 8 | 1511 | |||
GenBank mRNA | AF057286.1 | - | 6 | 966 | ||
AK036331.1 | - | 7 | 1449 | |||
AK139234.1 | - | 9 | 1707 | |||
AK139793.1 | - | 9 | 1642 | |||
AK160782.1 | - | 7 | 1476 | |||
AK173093.1 | - | 9 | 1695 | |||
BC039138.1 | - | 8 | 1475 | 2647379 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
