siRNA Details
Gene Symbol: Eps15l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289859.1 | NP_001276788.1 | 17 | 1760 | ||
NM_007944.3 | NP_031970.2 | 17 | 1853 | |||
GenBank mRNA | AK036662.1 | - | 17 | 1819 | ||
AK146781.1 | - | 17 | 1831 | |||
AK152602.1 | - | 17 | 1738 | |||
U29156.1 | - | 17 | 1840 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
