siRNA Details
Gene Symbol: Nr2f6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010150.2 | NP_034280.2 | 1, 2 | 970 | ||
GenBank mRNA | AK131941.1 | - | 1, 2 | 934 | ||
AK146779.1 | - | 1, 2 | 674 | |||
AK171552.1 | - | 1, 2 | 467 | |||
BC008138.1 | - | 1, 2 | 689 | 3582557 | ||
L25674.1 | - | 1, 2 | 402 | |||
X76654.1 | - | 1, 2 | 489 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
