siRNA Details
Gene Symbol: Erbb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010154.1 | NP_034284.1 | 20 | 2683 | ||
GenBank mRNA | AK144050.1 | - | 20 | 2684 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
