siRNA Details
Gene Symbol: Erbb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010154.1 | NP_034284.1 | 4 | 800 | ||
GenBank mRNA | AK144050.1 | - | 4 | 801 |

More Information
Other Species Targeted

Gene Ontology Categories:
