siRNA Details
Gene Symbol: Ces1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007954.4 | NP_031980.2 | 8, 9 | 1072 | ||
GenBank mRNA | AI527022.1 | - | 4, 5 | 435 | ||
AK050444.1 | - | 8, 9 | 1073 | |||
M57960.1 | - | 8, 9 | 1065 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
