siRNA Details
Gene Symbol: Ces1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133660.3 | NP_598421.1 | 12 | 1560 | ||
GenBank mRNA | AK144549.1 | - | 12 | 1545 | ||
BC019208.1 | - | 12 | 1491 | 5052893 | ||
S80191.1 | - | 12 | 1490 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
