siRNA Details
Gene Symbol: Ces3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159415.1 | NP_001152887.1 | 6 | 804 | ||
NM_144511.2 | NP_653094.2 | 6 | 804 | |||
GenBank mRNA | BC010812.1 | - | 6 | 794 | ||
BC019147.1 | - | 6 | 800 | 5052937 | ||
BC019926.1 | - | 6 | 795 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
