siRNA Details
Gene Symbol: Gnb1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081682.2 | NP_001075151.1 | 4 | 414 | ||
NM_001285491.1 | NP_001272420.1 | 4 | 414 | |||
NM_001285493.1 | NP_001272422.1 | 4 | 356 | |||
NM_001285494.1 | NP_001272423.1 | 4 | 356 | |||
NM_023120.5 | NP_075609.2 | 4 | 356 | |||
GenBank mRNA | AF301595.1 | - | 4 | 307 | ||
AK010489.1 | - | 4 | 314 | |||
AK016952.1 | - | 4 | 344 | |||
AK087954.1 | - | 4 | 413 | |||
AK158210.1 | - | 4 | 343 | |||
AK166195.1 | - | 4 | 340 | |||
AK171253.1 | - | 4 | 336 | |||
AK173227.1 | - | 4 | 339 | |||
BC024635.1 | - | 4 | 325 | 4193427 | ||
BC037676.1 | - | 3 | 242 | 3493788 | ||
BY328275.1 | - | 4 | 362 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
