siRNA Details
Gene Symbol: Esx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007957.2 | NP_031983.2 | 5 | 1222 | ||
GenBank mRNA | AF004211.1 | - | 4 | 1310 | ||
AF017734.1 | - | 4 | 939 | |||
AF017735.1 | - | 5 | 1022 | |||
AF085715.1 | - | 5 | 1223 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
