siRNA Details
Gene Symbol: Celf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110228.1 | NP_001103698.1 | 5 | 519 | ||
NM_001110229.1 | NP_001103699.1 | 6 | 606 | |||
NM_001110230.1 | NP_001103700.1 | 6 | 606 | |||
NM_001110231.1 | NP_001103701.1 | 8 | 581 | |||
NM_001110232.1 | NP_001103702.1 | 7 | 592 | |||
NM_001160292.1 | NP_001153764.1 | 6 | 606 | |||
NM_001160293.1 | NP_001153765.1 | 5 | 519 | |||
NM_010160.2 | NP_034290.2 | 5 | 519 | |||
GenBank mRNA | AF090696.1 | - | 7 | 592 | ||
AF090697.1 | - | 5 | 492 | |||
AK135166.1 | - | 5 | 470 | |||
AK137292.1 | - | 6 | 547 | |||
AK151818.1 | - | 4 | 377 | |||
AK152628.1 | - | 4 | 377 | |||
AK153267.1 | - | 4 | 377 | |||
AK159996.1 | - | 6 | 611 | |||
AK160861.1 | - | 8 | 582 | |||
AK162651.1 | - | 4 | 378 | |||
AK164914.1 | - | 6 | 547 | |||
AK172648.1 | - | 4 | 377 | |||
AK206137.1 | - | 4 | 205 | |||
BC026856.1 | - | 5 | 470 | 4503295 | ||
Y18298.1 | - | 4 | 385 |
