siRNA Details
Gene Symbol: Celf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160292.1 | NP_001153764.1 | 12, 13 | 1438 | ||
NM_010160.2 | NP_034290.2 | 11, 12 | 1351 | |||
GenBank mRNA | AK135166.1 | - | 11, 12 | 1303 | ||
AK152628.1 | - | 10, 11 | 1221 | |||
AK159996.1 | - | 12, 13 | 1443 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
