siRNA Details
Gene Symbol: Celf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110228.1 | NP_001103698.1 | 1 | 111 | ||
NM_001160293.1 | NP_001153765.1 | 1 | 111 | |||
NM_010160.2 | NP_034290.2 | 1 | 111 | |||
GenBank mRNA | AF090697.1 | - | 1 | 84 | ||
AK052102.1 | - | 1 | 44 | |||
AK135166.1 | - | 1 | 62 | |||
BC026856.1 | - | 1 | 62 | 4503295 | ||
BY145030.1 | - | 1 | 68 | |||
CF536042.1 | - | 1 | 111 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_017197, NM_001083586, XM_008771861, AF090695, AJ010386.
Gene Ontology Categories:
