siRNA Details
Gene Symbol: Evl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163394.1 | NP_001156866.1 | 4, 5 | 913 | ||
NM_001163395.1 | NP_001156867.1 | 4, 5 | 665 | |||
NM_001163396.2 | NP_001156868.1 | 5, 6 | 687 | |||
NM_007965.3 | NP_031991.3 | 4, 5 | 913 | |||
GenBank mRNA | AF279662.1 | - | 3, 4 | 424 | ||
AK134563.1 | - | 4, 5 | 665 | |||
AK137823.1 | - | 4, 5 | 502 | |||
AK151633.1 | - | 5, 6 | 688 | |||
BC059810.1 | - | 4, 5 | 913 | 6828523 | ||
BC096017.1 | - | 4, 5 | 806 | |||
U72519.1 | - | 4, 5 | 676 |

More Information
Other Species Targeted

Gene Ontology Categories:
