siRNA Details
Gene Symbol: Evx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007967.2 | NP_031993.1 | 1 | 213 | ||
GenBank mRNA | BC061467.1 | - | 351 | 6827284 | ||
M93128.1 | - | 1 | 213 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
