siRNA Details
Gene Symbol: Eya2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271962.1 | NP_001258891.1 | 11, 12 | 1084 | ||
NM_001271963.1 | NP_001258892.1 | 11, 12 | 1260 | |||
NM_010165.3 | NP_034295.1 | 11, 12 | 1261 | |||
GenBank mRNA | AK137718.1 | - | 11, 12 | 1256 | ||
AK141765.1 | - | 11, 12 | 1260 | |||
AY212511.1 | - | 8, 9 | 775 | |||
AY212512.1 | - | 9, 10 | 931 | |||
BC003755.1 | - | 11, 12 | 1261 | 3497245 | ||
U61111.1 | - | 10, 11 | 1002 | |||
U71208.1 | - | 11, 12 | 1086 | |||
U81603.1 | - | 10, 11 | 1001 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
