siRNA Details
Gene Symbol: Eya4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010167.4 | NP_034297.2 | 14 | 1467 | ||
GenBank mRNA | AK041625.1 | - | 14 | 1468 | ||
BC120898.1 | - | 14 | 1451 | 40061597 | ||
BC120899.1 | - | 14 | 1451 | 40061599 | ||
Y17115.1 | - | 15 | 1578 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
