siRNA Details
Gene Symbol: Ezh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB004817.1 | - | 7 | 757 | ||
AF483490.1 | - | 5 | 646 | |||
AF483491.1 | - | 5 | 646 | |||
AK045374.1 | - | 7 | 745 | |||
AK129004.1 | - | 6 | 717 | |||
AK138942.1 | - | 7 | 740 | |||
AK140694.1 | - | 7 | 764 | |||
AK164192.1 | - | 7 | 756 | |||
BC007135.1 | - | 6 | 717 | 3600884 | ||
U60453.1 | - | 7 | 745 |
