siRNA Details
Gene Symbol: Sfxn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027324.5 | NP_081600.1 | 2 | 210 | ||
GenBank mRNA | AF325260.1 | - | 2 | 188 | ||
AK012650.1 | - | 2 | 209 | |||
AK151110.1 | - | 2 | 156 | |||
BC005743.1 | - | 2 | 167 | 3599636 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
