siRNA Details
Gene Symbol: Fabp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007980.3 | NP_032006.1 | 3 | 670 | ||
GenBank mRNA | AK007995.1 | - | 3 | 368 | ||
AY523818.1 | - | 3 | 356 | |||
AY523819.1 | - | 3 | 356 | |||
BC013457.1 | - | 3 | 350 | 4218050 | ||
BY707992.1 | - | 3 | 366 | |||
M65034.1 | - | 3 | 364 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
