siRNA Details
Gene Symbol: Fadd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010175.5 | NP_034305.1 | 2 | 1610 | ||
GenBank mRNA | AK031396.1 | - | 2 | 1611 | ||
AK084808.1 | - | 2 | 1611 | |||
AK169798.1 | - | 2 | 1638 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
