siRNA Details
Gene Symbol: Fasn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007988.3 | NP_032014.3 | 42 | 7713 | ||
GenBank mRNA | AF127033.1 | - | 42 | 7230 | ||
AK080374.1 | - | 16 | 2610 | |||
AK147214.1 | - | 42 | 7409 | |||
AK147694.1 | - | 42 | 7477 | |||
BC010310.1 | - | 11 | 1700 | |||
BC046513.1 | - | 42 | 7267 | 4196969 | ||
BC059850.1 | - | 14 | 2245 | |||
X13135.1 | - | 14 | 2188 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
