siRNA Details
Gene Symbol: Fbl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007991.3 | NP_032017.2 | 6 | 708 | ||
GenBank mRNA | AK012620.1 | - | 6 | 704 | ||
AK146328.1 | - | 6 | 700 | |||
BC003813.1 | - | 6 | 720 | 3487295 | ||
BC092274.1 | - | 6 | 746 | 4196645 | ||
BU511282.1 | - | 6 | 715 | |||
Z22593.1 | - | 6 | 696 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
