siRNA Details
Gene Symbol: Fbn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007993.2 | NP_032019.2 | 65 | 8457 | ||
GenBank mRNA | AF007248.1 | - | 11123 | |||
AK137018.1 | - | 9 | 1308 | |||
AK164949.1 | - | 8 | 1113 | |||
BC003905.1 | - | 2 | 369 | |||
BC055932.1 | - | 2 | 193 | |||
L29454.1 | - | 10544 | ||||
U22493.1 | - | 65 | 8390 |
