siRNA Details
Gene Symbol: Fbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019395.3 | NP_062268.1 | 5 | 877 | ||
GenBank mRNA | AJ132693.1 | - | 5 | 828 | ||
AK002216.1 | - | 5 | 739 | |||
AK149527.1 | - | 5 | 758 | |||
BC011480.1 | - | 5 | 741 | 4194764 | ||
BC051392.1 | - | 5 | 741 | 5054854 | ||
Y11067.1 | - | 2 | 181 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
