siRNA Details
Gene Symbol: Fcgr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010186.5 | NP_034316.1 | 4 | 444 | ||
GenBank mRNA | AF143154.1 | - | 4 | 437 | ||
AF143155.1 | - | 4 | 437 | |||
AF143156.1 | - | 4 | 437 | |||
AF143157.1 | - | 4 | 437 | |||
AF143158.1 | - | 4 | 437 | |||
AF143159.1 | - | 4 | 437 | |||
AF143160.1 | - | 4 | 437 | |||
AF143161.1 | - | 4 | 437 | |||
AF143162.1 | - | 4 | 437 | |||
AF143163.1 | - | 4 | 437 | |||
AF143164.1 | - | 4 | 437 | |||
AF143165.1 | - | 4 | 437 | |||
AF143166.1 | - | 4 | 437 | |||
AF143167.1 | - | 4 | 437 | |||
AF143168.1 | - | 4 | 437 | |||
AF143169.1 | - | 4 | 437 | |||
AF143170.1 | - | 4 | 437 | |||
AF143171.1 | - | 4 | 437 | |||
AF143172.1 | - | 4 | 437 | |||
AF143173.1 | - | 4 | 437 | |||
AF143174.1 | - | 4 | 437 | |||
AF143175.1 | - | 4 | 437 | |||
AF143176.1 | - | 4 | 437 | |||
AF143177.1 | - | 4 | 437 | |||
AF143178.1 | - | 4 | 437 | |||
AF143179.1 | - | 4 | 437 | |||
AF143180.1 | - | 4 | 437 | |||
AF143186.1 | - | 4 | 437 | |||
AF143187.1 | - | 4 | 437 | |||
AF143188.1 | - | 4 | 437 | |||
AK033874.1 | - | 4 | 445 | |||
M31314.1 | - | 4 | 438 |
