siRNA Details
Gene Symbol: Fcgr2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077189.1 | NP_001070657.1 | 2, 3 | 192 | ||
NM_010187.2 | NP_034317.1 | 2, 3 | 192 | |||
GenBank mRNA | AK080885.1 | - | 2, 3 | 195 | ||
AK089266.1 | - | 2, 3 | 193 | |||
AK150613.1 | - | 2, 3 | 193 | |||
AK155329.1 | - | 2, 3 | 184 | |||
AK170035.1 | - | 2, 3 | 182 | |||
AK171254.1 | - | 2, 3 | 184 | |||
AK171316.1 | - | 2, 3 | 175 | |||
BC038070.1 | - | 2, 3 | 179 | 3993915 | ||
M14216.1 | - | 2, 3 | 150 | |||
M14276.1 | - | 2, 3 | 88 | |||
M16367.1 | - | 4, 5 | 433 | |||
M17515.1 | - | 2, 3 | 146 | |||
U51629.1 | - | 2, 3 | 100 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
