siRNA Details
Gene Symbol: Fcgr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK169887.1 | - | 5 | 720 | ||
AK171289.1 | - | 5 | 724 | |||
AY897421.1 | - | 5 | 712 | |||
AY897422.1 | - | 5 | 712 | |||
AY897423.1 | - | 5 | 712 | |||
AY897424.1 | - | 5 | 707 | |||
AY897425.1 | - | 5 | 717 | |||
AY897426.1 | - | 5 | 717 | |||
AY897427.1 | - | 5 | 717 | |||
AY897428.1 | - | 5 | 721 | |||
AY897429.1 | - | 5 | 712 | |||
M14215.1 | - | 5 | 758 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
