siRNA Details
Gene Symbol: Fcgr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK169887.1 | - | 4 | 472 | ||
AK171289.1 | - | 4 | 476 | |||
AY897421.1 | - | 4 | 464 | |||
AY897422.1 | - | 4 | 464 | |||
AY897423.1 | - | 4 | 464 | |||
AY897424.1 | - | 4 | 459 | |||
AY897425.1 | - | 4 | 469 | |||
AY897426.1 | - | 4 | 469 | |||
AY897427.1 | - | 4 | 469 | |||
AY897428.1 | - | 4 | 473 | |||
M14215.1 | - | 4 | 510 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
