siRNA Details
Gene Symbol: Fcnb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010190.1 | NP_034320.1 | 9 | 776 | ||
GenBank mRNA | AF063217.1 | - | 9 | 749 | ||
AK010913.1 | - | 8 | 775 | |||
BC107220.1 | - | 9 | 776 | 40051808 | ||
BC107221.1 | - | 9 | 776 | 40051810 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
