siRNA Details
Gene Symbol: Fem1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010193.4 | NP_034323.1 | 2 | 1352 | ||
GenBank mRNA | AB022863.1 | - | 2 | 1262 | ||
AF064448.1 | - | 2 | 1083 | |||
AF178633.1 | - | 2 | 1009 | |||
AK032338.1 | - | 2 | 1329 | |||
AK122272.1 | - | 1 | 550 | |||
AK132692.1 | - | 2 | 1345 | |||
AK145371.1 | - | 1 | 715 | |||
AK149329.1 | - | 2 | 1315 | |||
AK154060.1 | - | 2 | 1640 | |||
BC068236.1 | - | 2 | 1306 | 30544277 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001108157, XM_008758662.
Gene Ontology Categories:
