siRNA Details
Gene Symbol: Fgd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008001.4 | NP_032027.2 | 10 | 2324 | ||
GenBank mRNA | AK148151.1 | - | 10 | 2325 | ||
AK219664.1 | - | 1 | 125 | |||
BC004838.1 | - | 2 | 214 | |||
BC011462.1 | - | 10 | 2261 | 3674293 | ||
U22325.1 | - | 10 | 1923 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
