siRNA Details
Gene Symbol: Fgd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008001.4 | NP_032027.2 | 16 | 2818 | ||
GenBank mRNA | AK148151.1 | - | 16 | 2819 | ||
BC004838.1 | - | 8 | 708 | |||
BC011462.1 | - | 16 | 2755 | 3674293 | ||
U22325.1 | - | 16 | 2417 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
