siRNA Details
Gene Symbol: Fgf17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008004.4 | NP_032030.1 | 5 | 893 | ||
GenBank mRNA | AB009250.1 | - | 5 | 397 | ||
AK077555.1 | - | 5 | 895 | |||
AK082858.1 | - | 5 | 784 | |||
BC119033.1 | - | 5 | 879 | 8733782 | ||
BC144776.1 | - | 5 | 846 | 9053324 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
