siRNA Details
Gene Symbol: HLA-DQB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243962.1 | NP_001230891.1 | 1, 2 | 198 | ||
GenBank mRNA | AB209638.1 | - | 1, 2 | 178 | ||
AF113250.1 | - | 1 | 17 | |||
AF195245.1 | - | 1 | 23 | |||
AF291674.1 | - | 1, 2 | 119 | |||
AK295255.1 | - | 1, 2 | 166 | |||
AK295476.1 | - | 1, 2 | 171 | |||
AK295695.1 | - | 1, 2 | 169 | |||
AK296060.1 | - | 1, 2 | 163 | |||
AK296613.1 | - | 1, 2 | 180 | |||
AK304543.1 | - | 1, 2 | 205 | |||
AY656681.1 | - | 1, 2 | 119 | |||
AY656682.1 | - | 1, 2 | 119 | |||
BC012106.1 | - | 1, 2 | 173 | 4574913 | ||
DQ136036.1 | - | 1, 2 | 164 | |||
J00198.1 | - | 1 | 28 | |||
KC879155.1 | - | 11 | ||||
L34095.1 | - | 1 | 23 | |||
L34096.1 | - | 1 | 23 | |||
L34097.1 | - | 1 | 23 | |||
L34098.1 | - | 1 | 23 | |||
L34101.1 | - | 1 | 23 | |||
L34102.1 | - | 1 | 23 | |||
L34103.1 | - | 1 | 23 | |||
L34106.1 | - | 1 | 23 | |||
L34107.1 | - | 1 | 23 | |||
L40179.1 | - | 1 | 23 | |||
L40180.1 | - | 1 | 23 | |||
L42626.1 | - | 1 | 23 | |||
M14189.1 | - | 1 | 23 | |||
M16996.1 | - | 1, 2 | 119 | |||
M17206.1 | - | 1 | 23 | |||
M17563.1 | - | 1, 2 | 131 | |||
M17564.1 | - | 1, 2 | 131 | |||
M17565.1 | - | 1, 2 | 87 | |||
M17955.1 | - | 1, 2 | 191 | |||
M19239.1 | - | 1 | 23 | |||
M20505.1 | - | 1, 2 | 87 | |||
M21135.1 | - | 1 | 23 | |||
M24364.1 | - | 1, 2 | 163 | |||
M25325.1 | - | 1, 2 | 201 | |||
M25326.1 | - | 1, 2 | 161 | |||
M32577.1 | - | 1, 2 | 87 | |||
M35982.1 | - | 1, 2 | 102 | |||
M59800.1 | - | 1 | 23 | |||
M59801.1 | - | 1 | 23 | |||
M60028.1 | - | 1, 2 | 176 | |||
M81140.1 | - | 1, 2 | 140 | |||
M81141.1 | - | 1, 2 | 154 | |||
M83770.1 | - | 1 | 23 | |||
S79787.1 | - | 11 | ||||
U63321.1 | - | 1 | 17 | |||
U66400.1 | - | 1 | 23 | |||
U83582.1 | - | 1, 2 | 117 | |||
U83585.1 | - | 1, 2 | 117 | |||
V00529.1 | - | 1 | 28 | |||
X00369.1 | - | 1, 2 | 87 | |||
X03068.1 | - | 1, 2 | 200 | |||
X76553.1 | - | 1 | 23 | |||
X76554.1 | - | 1 | 23 | |||
X96420.1 | - | 1, 2 | 119 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
