siRNA Details
Gene Symbol: Fgf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010204.1 | NP_034334.1 | 3 | 591 | ||
GenBank mRNA | AK086530.1 | - | 3 | 509 | ||
M92416.1 | - | 3 | 493 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
