siRNA Details
Gene Symbol: Fgfbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271616.1 | NP_001258545.1 | 3 | 491 | ||
NM_008009.4 | NP_032035.2 | 2 | 441 | |||
GenBank mRNA | AF065441.1 | - | 1 | 334 | ||
AK146625.1 | - | 2 | 406 | |||
BC065774.1 | - | 2 | 404 | 3975150 | ||
DV059653.1 | - | 3 | 473 | |||
U49641.1 | - | 1 | 1439 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
