siRNA Details
Gene Symbol: Fgfr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079908.2 | NP_001073377.1 | 6 | 1450 | ||
NM_001079909.2 | NP_001073378.1 | 5 | 1189 | |||
NM_010206.3 | NP_034336.2 | 6 | 1456 | |||
GenBank mRNA | AF176552.1 | - | 4 | 481 | ||
AK028354.1 | - | 5 | 629 | |||
AK049704.1 | - | 6 | 1416 | |||
BC010200.1 | - | 6 | 1349 | 3967874 | ||
BC033447.1 | - | 4 | 452 | 4037899 | ||
M28998.1 | - | 5 | 754 | |||
M33760.1 | - | 4 | 473 | |||
M65053.1 | - | 5 | 727 | |||
U22324.1 | - | 5 | 754 | |||
U23445.1 | - | 4 | 487 | |||
X51893.1 | - | 6 | 811 | |||
X57790.1 | - | 5 | 691 |
