siRNA Details
Gene Symbol: Fgfr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163215.2 | NP_001156687.1 | 10 | 1600 | ||
NM_001163216.2 | NP_001156688.1 | 8 | 1315 | |||
NM_001163217.2 | NP_001156689.1 | 9 | 1375 | |||
NM_001205270.1 | NP_001192199.1 | 10 | 1577 | |||
NM_008010.5 | NP_032036.2 | 10 | 1574 | |||
GenBank mRNA | AF024638.1 | - | 8 | 1223 | ||
AK143592.1 | - | 10 | 1510 | |||
AK163495.1 | - | 8 | 1153 | |||
BC053056.1 | - | 10 | 1484 | 5708838 | ||
BU053488.1 | - | 5 | 590 | |||
M81342.1 | - | 10 | 1492 | |||
S56291.1 | - | 9 | 1319 | |||
X58255.1 | - | 10 | 1478 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
