siRNA Details
Gene Symbol: HLA-DQB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243962.1 | NP_001230891.1 | 2 | 269 | ||
GenBank mRNA | AK295255.1 | - | 2 | 237 | ||
AK296060.1 | - | 2 | 234 | |||
AK304543.1 | - | 2 | 276 | |||
J00198.1 | - | 1 | 99 | |||
L34095.1 | - | 1 | 94 | |||
L40179.1 | - | 1 | 94 | |||
M14189.1 | - | 1 | 94 | |||
M81140.1 | - | 2 | 211 | |||
M81141.1 | - | 2 | 225 | |||
S79787.1 | - | 1 | 82 | |||
V00529.1 | - | 1 | 99 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
