siRNA Details
Gene Symbol: Fgfr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163215.2 | NP_001156687.1 | 15, 16 | 2346 | ||
NM_001163216.2 | NP_001156688.1 | 13, 14 | 2061 | |||
NM_001163217.2 | NP_001156689.1 | 14, 15 | 2121 | |||
NM_001205270.1 | NP_001192199.1 | 15, 16 | 2323 | |||
NM_008010.5 | NP_032036.2 | 15, 16 | 2320 | |||
GenBank mRNA | AF024638.1 | - | 13, 14 | 1969 | ||
AK143592.1 | - | 15, 16 | 2256 | |||
AK163495.1 | - | 13, 14 | 1899 | |||
BC053056.1 | - | 15, 16 | 2230 | 5708838 | ||
M81342.1 | - | 15, 16 | 2238 | |||
S56291.1 | - | 14, 15 | 2065 | |||
X58255.1 | - | 15, 16 | 2224 |
