siRNA Details
Gene Symbol: Ctgf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010217.2 | NP_034347.2 | 4, 5 | 1035 | ||
GenBank mRNA | BC006783.1 | - | 4, 5 | 956 | 3589136 | |
BQ892143.1 | - | 1, 2 | 172 | |||
M70642.1 | - | 4, 5 | 888 | |||
M80263.1 | - | 4, 5 | 954 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
