siRNA Details
Gene Symbol: Fkbp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB241120.1 | - | 4 | 375 | ||
AB241121.1 | - | 4 | 268 | |||
AF483488.1 | - | 3 | 140 | |||
AF483489.1 | - | 3 | 140 | |||
AK002777.1 | - | 3 | 247 | |||
AK010693.1 | - | 3 | 271 | |||
AK019362.1 | - | 3 | 275 | |||
AK145400.1 | - | 3 | 264 | |||
AK145986.1 | - | 2 | 297 | |||
AK151047.1 | - | 1 | 58 | |||
AK154751.1 | - | 3 | 219 | |||
AK168333.1 | - | 3 | 255 | |||
AK169186.1 | - | 3 | 178 | |||
AK169242.1 | - | 3 | 262 | |||
BC004671.1 | - | 3 | 222 | 3498682 | ||
BY065108.1 | - | 3 | 349 | |||
CX239711.1 | - | 4 | 355 | |||
X60203.1 | - | 3 | 237 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
