siRNA Details
Gene Symbol: Fkbp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010220.4 | NP_034350.1 | 5, 6 | 721 | ||
GenBank mRNA | AK169795.1 | - | 5, 6 | 662 | ||
BC015260.1 | - | 5, 6 | 660 | 4237766 | ||
BU613862.1 | - | 3, 4 | 388 | |||
CA317455.1 | - | 5, 6 | 684 | |||
CT010372.1 | - | 4, 5 | 502 | |||
U16959.1 | - | 5, 6 | 721 | |||
U36220.1 | - | 5, 6 | 649 |
