siRNA Details
Gene Symbol: HLA-DRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK289463.1 | - | 2 | 236 | ||
M11161.1 | - | 2 | 173 | |||
M21008.1 | - | 1 | 66 | |||
M33600.1 | - | 2 | 243 | |||
X03069.1 | - | 2 | 235 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
