siRNA Details
Gene Symbol: HLA-DRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK289463.1 | - | 2 | 187 | ||
DQ002917.1 | - | 2 | 124 | |||
M11161.1 | - | 2 | 124 | |||
M21008.1 | - | 1 | 17 | |||
M33600.1 | - | 2 | 194 | |||
X03069.1 | - | 2 | 186 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
