siRNA Details
Gene Symbol: HLA-DRB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021983.4 | NP_068818.4 | 4 | 737 | ||
GenBank mRNA | AF048707.1 | - | 3 | 527 | ||
AF361548.1 | - | 5 | 675 | |||
AF361549.1 | - | 5 | 675 | |||
AK225683.1 | - | 4 | 729 | |||
AK225690.1 | - | 4 | 729 | |||
AK291499.1 | - | 4 | 729 | |||
AK292151.1 | - | 5 | 717 | |||
AK293020.1 | - | 4 | 770 | |||
AY961064.1 | - | 4 | 666 | |||
AY961067.1 | - | 4 | 666 | |||
AY961070.2 | - | 4 | 666 | |||
BC005312.1 | - | 5 | 729 | 3933019 | ||
DQ284431.1 | - | 4 | 727 | |||
DQ284433.1 | - | 4 | 728 | |||
GQ891550.1 | - | 4 | 666 | |||
M15178.1 | - | 5 | 608 | |||
M16942.1 | - | 5 | 694 | |||
M17385.1 | - | 3 | 579 |
Gene Symbol: HLA-DRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243965.1 | NP_001230894.1 | 4 | 760 | ||
NM_002124.3 | NP_002115.2 | 4 | 760 | |||
GenBank mRNA | AF291675.1 | - | 4 | 666 | ||
AJ297583.1 | - | 4 | 666 | |||
AJ297585.1 | - | 4 | 666 | |||
AJ297586.2 | - | 4 | 666 | |||
AJ297587.1 | - | 4 | 666 | |||
AJ697892.1 | - | 4 | 675 | |||
AJ697893.1 | - | 4 | 675 | |||
AK289463.1 | - | 4 | 729 | |||
AK291987.1 | - | 4 | 760 | |||
AY626551.1 | - | 4 | 666 | |||
AY770514.1 | - | 4 | 666 | |||
AY935719.1 | - | 4 | 666 | |||
AY961062.1 | - | 4 | 666 | |||
AY961063.1 | - | 4 | 666 | |||
AY961065.1 | - | 4 | 666 | |||
AY961066.1 | - | 4 | 666 | |||
AY961068.1 | - | 4 | 666 | |||
AY961072.1 | - | 4 | 666 | |||
AY961073.1 | - | 4 | 666 | |||
AY961076.1 | - | 4 | 666 | |||
AY961077.1 | - | 4 | 666 | |||
AY961078.1 | - | 4 | 666 | |||
BC007920.2 | - | 4 | 712 | 4309837 | ||
BC008403.1 | - | 4 | 729 | 4272571 | ||
BC024269.1 | - | 4 | 709 | 4852401 | ||
BC031023.1 | - | 4 | 729 | 4695321 | ||
BC033827.1 | - | 4 | 709 | 5216243 | ||
BC108922.1 | - | 4 | 721 | 40023604 | ||
DQ002917.1 | - | 4 | 666 | |||
DQ090958.1 | - | 4 | 666 | |||
DQ140279.1 | - | 4 | 726 | |||
HG974574.1 | - | 4 | 666 | |||
HG974596.1 | - | 4 | 666 | |||
HG974600.1 | - | 4 | 666 | |||
HM067861.1 | - | 4 | 666 | |||
HM067862.1 | - | 4 | 666 | |||
HM067863.1 | - | 4 | 666 | |||
KJ534866.1 | - | 4 | 666 | |||
L02545.1 | - | 4 | 680 | |||
L42143.1 | - | 4 | 666 | |||
L76133.1 | - | 4 | 706 | |||
M11161.1 | - | 4 | 666 | |||
M11867.1 | - | 4 | 695 | |||
M14661.1 | - | 3 | 528 | |||
M14662.1 | - | 3 | 579 | |||
M16957.1 | - | 3 | 579 | |||
M16959.1 | - | 3 | 579 | |||
M17378.1 | - | 3 | 579 | |||
M17381.1 | - | 3 | 579 | |||
M17384.1 | - | 3 | 579 | |||
M20138.1 | - | 4 | 668 | |||
M20430.1 | - | 4 | 709 | |||
M20504.1 | - | 5 | 701 | |||
M21008.1 | - | 3 | 559 | |||
M26038.1 | - | 4 | 712 | |||
M27635.1 | - | 4 | 703 | |||
M28583.1 | - | 5 | 705 | |||
M28584.1 | - | 4 | 637 | |||
M33600.1 | - | 4 | 736 | |||
U56640.1 | - | 3 | 523 | |||
U65585.1 | - | 4 | 706 | |||
U95817.1 | - | 3 | 579 | |||
X00699.1 | - | 4 | 701 | |||
X00700.1 | - | 4 | 670 | |||
X02902.1 | - | 4 | 722 | |||
X03069.1 | - | 4 | 728 |
Gene Symbol: HLA-DRB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022555.3 | NP_072049.2 | 4 | 706 | ||
GenBank mRNA | A06800.1 | - | 4 | 702 | ||
AF000448.1 | - | 4 | 579 | |||
AF192258.1 | - | 3 | 579 | |||
AF192259.1 | - | 3 | 579 | |||
AF199236.2 | - | 5 | 687 | |||
AK226043.1 | - | 4 | 732 | |||
AY138123.1 | - | 4 | 666 | |||
BC001023.2 | - | 4 | 726 | 3345335 | ||
BC008987.1 | - | 4 | 690 | 4184806 | ||
BC106057.1 | - | 4 | 769 | 4385464 | ||
KJ534865.1 | - | 4 | 666 | |||
M16941.1 | - | 4 | 689 | |||
M17380.1 | - | 3 | 579 | |||
M28991.1 | - | 4 | 624 | |||
M28992.1 | - | 4 | 585 | |||
U66825.1 | - | 4 | 706 | |||
U95819.1 | - | 4 | 680 | |||
V00522.1 | - | 4 | 701 |
Gene Symbol: HLA-DRB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB062112.2 | - | 4 | 710 | ||
AF490771.1 | - | 4 | 647 | |||
AJ297582.1 | - | 4 | 666 | |||
AK292140.1 | - | 4 | 729 | |||
M17379.1 | - | 3 | 579 | |||
M17382.1 | - | 3 | 579 | |||
M17383.1 | - | 3 | 579 | |||
M27689.1 | - | 3 | 579 | |||
U66826.1 | - | 4 | 706 | |||
X99895.1 | - | 4 | 673 |
Gene Symbol: LOC105369230 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | HG974601.1 | - | 4 | 666 | ||
HM067848.1 | - | 4 | 666 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
