siRNA Details
Gene Symbol: Foxl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008024.2 | NP_032050.2 | 1 | 692 | ||
GenBank mRNA | AB221616.1 | - | 1 | 415 | ||
BC137805.1 | - | 1 | 437 | 8860824 | ||
BC137806.1 | - | 1 | 437 | 8860825 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
