siRNA Details
Gene Symbol: Fmo5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161763.1 | NP_001155235.1 | 9 | 1650 | ||
NM_001161765.1 | NP_001155237.1 | 8 | 1733 | |||
NM_010232.4 | NP_034362.2 | 9 | 1634 | |||
GenBank mRNA | AK133675.1 | - | 8 | 1715 | ||
AK137645.1 | - | 5 | 916 | |||
AK147040.1 | - | 9 | 1627 | |||
BC022991.1 | - | 9 | 1597 | 5123876 |
