siRNA Details
Gene Symbol: Fosb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008036.2 | NP_032062.1 | 4 | 1523 | ||
GenBank mRNA | BC132064.1 | - | 4 | 1078 | 40130341 | |
BC137994.1 | - | 4 | 1078 | 8861013 | ||
BC145082.1 | - | 5 | 938 | |||
CK795975.1 | - | 3 | 624 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
